(5-Fluoro-[1,1'-biphenyl]-3-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

67470-500MG 67470-1G 67470-5G
77335-896EA 367.71 USD
77335-896 77357-156 77381-064
(5-Fluoro-[1,1'-biphenyl]-3-yl)boronic acid ≥95%
(5-Fluoro-[1,1'-biphenyl]-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BFO₂
MW: 216.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28167220
CAS Number: 1315280-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR