1,5-Diethoxy-2,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

67460-500MG 67460-1G 67460-5G 67460-25G
77332-894EA 182.31 USD
77332-894 77346-894 77368-936 77390-084
1,5-Diethoxy-2,4-difluorobenzene ≥97%
1,5-Diethoxy-2,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34762822
CAS Number: 2901095-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR