5-Ethoxy-2,4-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

67459-500MG 67459-1G 67459-5G 67459-25G
77332-892EA 187.46 USD
77332-892 77357-154 77368-934 77390-082
5-Ethoxy-2,4-difluorophenol ≥97%
5-Ethoxy-2,4-difluorophenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29044505
CAS Number: 1881295-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR