2-Fluoro-3-(trimethylsilyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

67456-1G 67456-5G 67456-25G
77341-216EA 76.22 USD
77341-216 77368-932 77381-062
2-Fluoro-3-(trimethylsilyl)pyridine ≥97%
2-Fluoro-3-(trimethylsilyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₂FNSi
MW: 169.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18803376
CAS Number: 78498-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR