1-(3-Bromo-2-fluoro-6-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67452-500MG 67452-1G 67452-5G 67452-25G
77332-890EA 161.71 USD
77332-890 77345-960 77368-930 77390-080
1-(3-Bromo-2-fluoro-6-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥95%
1-(3-Bromo-2-fluoro-6-(trifluoromethyl)phenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₃BrF₆O
MW: 321.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762821
CAS Number: 2807467-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR