1-(3-Bromo-2,6-dichlorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67451-500MG 67451-1G 67451-5G 67451-25G
77332-888EA 95.79 USD
77332-888 77342-586 77368-928 77381-060
1-(3-Bromo-2,6-dichlorophenyl)-2,2-difluoroethanone ≥97%
1-(3-Bromo-2,6-dichlorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃BrCl₂F₂O
MW: 303.916 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31706959
CAS Number: 2807467-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR