1-(5-Chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67447-500MG 67447-1G 67447-25G 67447-5G
77332-886EA 293.55 USD
77332-886 77357-152 77390-078 77381-058
1-(5-Chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
1-(5-Chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762818
CAS Number: 2807469-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR