1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
67446-500MG
67446-1G
67446-5G
67446-25G
77332-884EA
116.39
USD
77332-884
77343-502
77368-926
77381-056
1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-difluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₆ClF₃O MW: 222.592 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32869200 CAS Number: 2807474-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...