1-(3-Ethoxy-2-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67443-500MG 67443-1G 67443-5G 67443-25G
77332-882EA 141.11 USD
77332-882 77344-878 77368-924 77390-076
1-(3-Ethoxy-2-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(3-Ethoxy-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762817
CAS Number: 2807474-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR