1-(6-Bromo-2,4-difluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
67426-500MG
67426-1G
67426-5G
67426-25G
77332-878EA
116.39
USD
77332-878
77343-452
77368-922
77381-052
1-(6-Bromo-2,4-difluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
1-(6-Bromo-2,4-difluoro-3-methylphenyl)-2,2-difluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₅BrF₄O MW: 285.033 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762810 CAS Number: 2807448-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...