2,2-Difluoro-1-(2-fluoro-5-iodophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

67424-500MG 67424-1G 67424-5G 67424-25G
77332-876EA 182.31 USD
77332-876 77347-008 77368-920 77390-074
2,2-Difluoro-1-(2-fluoro-5-iodophenyl)ethanone ≥95%
2,2-Difluoro-1-(2-fluoro-5-iodophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₄F₃IO
MW: 300.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31543048
CAS Number: 2354099-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR