1-(3-Bromo-2-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67423-500MG 67423-1G 67423-5G 67423-25G
77332-874EA 232.78 USD
77332-874 77357-148 77368-918 77390-072
1-(3-Bromo-2-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(3-Bromo-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
MW: 253.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762808
CAS Number: 1697801-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR