1-(5-Chloro-2,3,4-trifluorophenyl)-2,2-difluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
67411-500MG
67411-1G
67411-5G
67411-25G
77332-872EA
101.97
USD
77332-872
77343-006
77368-916
77381-050
1-(5-Chloro-2,3,4-trifluorophenyl)-2,2-difluoroethanone ≥95%
1-(5-Chloro-2,3,4-trifluorophenyl)-2,2-difluoroethanone
Precaución: for Research Use Only
Formula:
C₈H₂ClF₅O MW: 244.546 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762803 CAS Number: 2807449-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...