1-(2,6-Difluoro-3-methylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67402-500MG 67402-1G 67402-5G 67402-25G
77332-868EA 264.71 USD
77332-868 77357-146 77368-912 77390-068
1-(2,6-Difluoro-3-methylphenyl)-2,2-difluoroethanone ≥97%
1-(2,6-Difluoro-3-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆F₄O
MW: 206.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045309
CAS Number: 1565517-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR