2,2-Difluoro-1-(2-fluoro-3-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

67399-500MG 67399-1G 67399-5G 67399-25G
77332-866EA 223.51 USD
77332-866 77357-144 77368-910 77390-066
2,2-Difluoro-1-(2-fluoro-3-methoxyphenyl)ethanone ≥97%
2,2-Difluoro-1-(2-fluoro-3-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045342
CAS Number: 1566615-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR