2,2-Difluoro-1-(2-fluoro-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

67397-500MG 67397-1G 67397-5G
77339-142EA 554.14 USD
77339-142 77357-142 77381-048
2,2-Difluoro-1-(2-fluoro-5-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-fluoro-5-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O
MW: 188.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045355
CAS Number: 1564979-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR