(4-(Difluoromethyl)-3-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

67325-500MG 67325-1G 67325-5G
77332-858EA 340.93 USD
77332-858 77357-130 77381-040
(4-(Difluoromethyl)-3-fluorophenyl)boronic acid ≥97%
(4-(Difluoromethyl)-3-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₂
MW: 189.928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383465
CAS Number: 1254118-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR