2,2,2-Trifluoro-1-(2-fluoro-6-iodo-3-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
67286-500MG
67286-1G
67286-5G
67286-25G
77332-856EA
88.58
USD
77332-856
77342-334
77368-902
77381-038
2,2,2-Trifluoro-1-(2-fluoro-6-iodo-3-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-fluoro-6-iodo-3-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₅F₄IO₂ MW: 348.033 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29040798 CAS Number: 2091756-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...