1-(3-(Benzyloxy)-4-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67242-500MG 67242-1G 67242-5G 67242-25G
77332-854EA 140.08 USD
77332-854 77344-728 77368-900 77390-058
1-(3-(Benzyloxy)-4-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-(Benzyloxy)-4-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₅H₉ClF₄O₂
MW: 332.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30717185
CAS Number: 2386368-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR