1-(3,5-Dichloro-4-propoxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67042-500MG 67042-1G 67042-5G 67042-25G
77332-850EA 176.13 USD
77332-850 77346-562 77368-896 77390-056
1-(3,5-Dichloro-4-propoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(3,5-Dichloro-4-propoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₁H₉Cl₂F₃O₂
MW: 301.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762769
CAS Number: 1555134-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR