1-(3,5-Dichloro-4-ethoxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

66926-500MG 66926-1G 66926-5G 66926-25G
77332-844EA 280.16 USD
77332-844 77357-124 77368-892 77390-052
1-(3,5-Dichloro-4-ethoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(3,5-Dichloro-4-ethoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₇Cl₂F₃O₂
MW: 287.063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762768
CAS Number: 1557295-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR