1-(4-Bromo-2,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

66878-500MG 66878-1G 66878-5G 66878-25G
77332-842EA 161.71 USD
77332-842 77345-958 77368-890 77390-050
1-(4-Bromo-2,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-Bromo-2,5-dimethylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O
MW: 281.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107307
CAS Number: 1506273-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR