1-(2-Chloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
66845-500MG
66845-1G
66845-5G
77336-932EA
400.67
USD
77336-932
77357-118
77381-024
1-(2-Chloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Chloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₉H₃ClF₆O MW: 276.563 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28105426 CAS Number: 2173145-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...