1-(3-(Benzyloxy)-6-chloro-2-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

66782-500MG 66782-1G 66782-5G
77332-838EA 320.33 USD
77332-838 77357-112 77381-014
1-(3-(Benzyloxy)-6-chloro-2-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(3-(Benzyloxy)-6-chloro-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₀ClF₃O₂
MW: 314.687 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762763
CAS Number: 2807474-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR