1-(2,6-Difluoro-3-(methylthio)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

66778-500MG 66778-1G 66778-5G 66778-25G
77332-834EA 250.29 USD
77332-834 77357-108 77368-884 77390-046
1-(2,6-Difluoro-3-(methylthio)phenyl)-2,2-difluoroethanone ≥95%
1-(2,6-Difluoro-3-(methylthio)phenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆F₄OS
Storage Temperature: Ambient
MDL Number: MFCD34762760
CAS Number: 2807445-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR