2-Hydroxy-5-isobutylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

66759-500MG 66759-1G 66759-25G 66759-5G
77332-830EA 284.28 USD
77332-830 77357-104 77390-042 77381-010
2-Hydroxy-5-isobutylbenzoic acid ≥97%
2-Hydroxy-5-isobutylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27940775
CAS Number: 100245-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR