2-Bromo-4-chloro-1,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

66749-500MG 66749-1G 66749-5G
77340-352EA 916.7 USD
77340-352 77357-102 77381-008
2-Bromo-4-chloro-1,3-dimethoxybenzene ≥97%
2-Bromo-4-chloro-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO₂
MW: 251.505 g/mol
Storage Temperature: Ambient
CAS Number: 1597673-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR