1-Chloro-3,4-difluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

66739-500MG 66739-1G 66739-25G 66739-5G
77332-828EA 304.88 USD
77332-828 77357-100 77390-040 77381-006
1-Chloro-3,4-difluoro-2-iodobenzene ≥97%
1-Chloro-3,4-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂I
MW: 274.434 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922586
CAS Number: 1208074-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR