3-Fluoro-4-iodo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

66722-500MG 66722-1G 66722-5G 66722-25G
77332-826EA 103 USD
77332-826 77343-058 77368-880 77381-004
3-Fluoro-4-iodo-5-methoxybenzaldehyde ≥97%
3-Fluoro-4-iodo-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762747
CAS Number: 2090466-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR