4-(Benzyloxy)-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

66678-500MG 66678-1G 66678-5G 66678-25G
77332-822EA 178.19 USD
77332-822 77346-680 77368-876 77390-036
4-(Benzyloxy)-2,5-dimethylbenzoic acid ≥97%
4-(Benzyloxy)-2,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01929046
CAS Number: 354551-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR