(2,4-Dichlorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

66675-500MG 66675-1G 66675-5G 66675-25G
77332-820EA 202.91 USD
77332-820 77357-096 77368-874 77390-034
(2,4-Dichlorophenyl)trimethylsilane ≥95%
(2,4-Dichlorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₂Cl₂Si
Storage Temperature: Ambient
MDL Number: MFCD34687687
CAS Number: 149021-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR