4-Chloro-3,5-difluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

66649-500MG 66649-1G 66649-5G 66649-25G
77332-818EA 121.54 USD
77332-818 77343-872 77368-872 77381-000
4-Chloro-3,5-difluoro-N-methoxy-N-methylbenzamide ≥97%
4-Chloro-3,5-difluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈ClF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34762709
CAS Number: 1189175-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR