(3-Chloro-4-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

66603-500MG 66603-1G 66603-5G 66603-25G
77332-816EA 220.42 USD
77332-816 77357-092 77368-870 77390-032
(3-Chloro-4-fluorophenyl)trimethylsilane ≥95%
(3-Chloro-4-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₂ClFSi
MW: 202.728 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201827
CAS Number: 845823-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR