2-Chloro-1-fluoro-3-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

66592-500MG 66592-1G 66592-5G
77339-410EA 586.07 USD
77339-410 77357-090 77380-998
2-Chloro-1-fluoro-3-iodo-5-methylbenzene ≥97%
2-Chloro-1-fluoro-3-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751672
CAS Number: 1805639-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR