4'-Chloro-2',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

66579-500MG 66579-1G 66579-5G 66579-25G
77332-814EA 202.91 USD
77332-814 77357-088 77368-868 77390-030
4'-Chloro-2',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Chloro-2',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO
MW: 244.716 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19096720
CAS Number: 1409684-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR