N,2,3,4-Tetramethoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

66572-1G 66572-5G 66572-25G
77341-196EA 74.16 USD
77341-196 77368-864 77380-994
N,2,3,4-Tetramethoxy-N-methylbenzamide ≥97%
N,2,3,4-Tetramethoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD15376399
CAS Number: 1180287-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR