2,3,4-Trimethoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

66567-500MG 66567-1G 66567-5G
77337-988EA 456.29 USD
77337-988 77357-086 77380-992
2,3,4-Trimethoxy-N-methylbenzamide ≥97%
2,3,4-Trimethoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD08891826
CAS Number: 65489-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR