Methyl 5-acetyl-2-chlorobenzoate ≥95%

Supplier: AOBChem USA

Warning

66559-500MG 66559-1G 66559-5G
77337-546EA 419.21 USD
77337-546 77357-084 77380-990
Methyl 5-acetyl-2-chlorobenzoate ≥95%
Methyl 5-acetyl-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD20482193
CAS Number: 792911-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR