5-Acetyl-2-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

66558-500MG 66558-1G 66558-5G 66558-25G
77332-812EA 260.59 USD
77332-812 77357-082 77368-862 77390-028
5-Acetyl-2-chlorobenzoic acid ≥97%
5-Acetyl-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MW: 198.603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20644290
CAS Number: 98588-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR