3,5-Dichloro-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66554-500MG 66554-1G 66554-5G 66554-25G
77332-808EA 130.81 USD
77332-808 77344-352 77368-858 77390-024
3,5-Dichloro-2-methyl-1,1'-biphenyl ≥97%
3,5-Dichloro-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀Cl₂
Storage Temperature: Ambient
MDL Number: MFCD34762658
CAS Number: 2901084-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR