3-Chloro-2-fluoro-6-iodobenzamide ≥95%

Supplier: AOBChem USA

Warning

66507-500MG 66507-1G 66507-5G 66507-25G
77332-804EA 198.79 USD
77332-804 77357-076 77368-854 77390-020
3-Chloro-2-fluoro-6-iodobenzamide ≥95%
3-Chloro-2-fluoro-6-iodobenzamide

Precaución: for Research Use Only

Formula: C₇H₄ClFINO
Storage Temperature: Ambient
MDL Number: MFCD34762623
CAS Number: 1252989-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR