1,5-Dibromo-2-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

66505-1G 66505-5G 66505-25G
77340-802EA 60.77 USD
77340-802 77368-852 77380-986
1,5-Dibromo-2-methoxy-3-methylbenzene ≥97%
1,5-Dibromo-2-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20501464
CAS Number: 7463-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR