2-Fluoro-3-methyl-4-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

66496-500MG 66496-1G 66496-5G
77339-560EA 596.37 USD
77339-560 77357-074 77380-984
2-Fluoro-3-methyl-4-(methylthio)benzoic acid ≥97%
2-Fluoro-3-methyl-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD34762615
CAS Number: 2092362-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR