(3-Fluoro-4-iodo-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

66493-500MG 66493-1G 66493-5G
77338-054EA 460.41 USD
77338-054 77357-072 77380-982
(3-Fluoro-4-iodo-2-methylphenyl)(methyl)sulfane ≥97%
(3-Fluoro-4-iodo-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIS
Storage Temperature: Ambient
MDL Number: MFCD34762614
CAS Number: 2385562-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR