(4-Chloro-2-methylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

66450-500MG 66450-1G 66450-5G 66450-25G
77332-794EA 183.34 USD
77332-794 77347-130 77368-842 77390-016
(4-Chloro-2-methylphenyl)trimethylsilane ≥97%
(4-Chloro-2-methylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₅ClSi
MW: 198.765 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521277
CAS Number: 1314929-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR