3-Hydroxy-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

66448-500MG 66448-1G 66448-5G
77340-180EA 817.82 USD
77340-180 77357-066 77380-974
3-Hydroxy-5-(trifluoromethoxy)benzoic acid ≥97%
3-Hydroxy-5-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₄
MW: 222.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397463
CAS Number: 1163141-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR