3-Ethoxy-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

66434-500MG 66434-1G 66434-5G
77332-790EA 347.11 USD
77332-790 77357-064 77380-972
3-Ethoxy-5-(trifluoromethoxy)benzoic acid ≥97%
3-Ethoxy-5-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₄
MW: 250.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762589
CAS Number: 2807447-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR