3,4-Dichloro-2-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

66370-500MG 66370-1G
77340-506EA 1335.91 USD
77340-506 77357-058
3,4-Dichloro-2-ethoxybenzoic acid ≥97%
3,4-Dichloro-2-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34762538
CAS Number: 2385126-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR