3,4-Diethoxy-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

66351-500MG 66351-1G 66351-5G
77338-086EA 462.47 USD
77338-086 77357-052 77380-966
3,4-Diethoxy-5-fluorobenzoic acid ≥97%
3,4-Diethoxy-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD34762526
CAS Number: 2807468-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR