2-(Benzyloxy)-5-bromo-1-ethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

66342-500MG 66342-1G 66342-5G
77338-952EA 535.6 USD
77338-952 77357-050 77380-964
2-(Benzyloxy)-5-bromo-1-ethoxy-3-fluorobenzene ≥97%
2-(Benzyloxy)-5-bromo-1-ethoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrFO₂
MW: 325.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762518
CAS Number: 2807471-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR